Docetaxel ≥98.0%
Supplier: TCI America
Synonyms:
Taxotere
CAS Number: 114977-28-5
MDL Number: MFCD00800737
Molecular Formula: C43H53NO14
Molecular Weight: 807.89
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Regenerative Medicine Research Reagents, Antitumor Ingredients, Terpenoids (Phytochemicals), Diterpenes, Cancer Research Reagents
Other Product Names / Synonyms: N-Debenzoyl-N-tert-butoxycarbonyl-10-deacetyltaxol
Formula:
C₄₃H₅₃NO₁₄ MW: 807.89 g/mol Melting Pt: 188 °C |
MDL Number:
MFCD00800737 CAS Number: 114977-28-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...