1,2-Dimyristoyl-sn-glycero-3-phosphocholine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 14:0 PC, 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, DMPC, β-γ-Dimyristoyl-L-α-Lecithin, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate

D3924-1G D3924-200MG
TCD3924-1GEA 335.8 USD
TCD3924-1G TCD3924-200MG
1,2-Dimyristoyl-sn-glycero-3-phosphocholine ≥97.0% (by HPLC, titration analysis)
1,2-Dimyristoyl-sn-glycero-3-phosphocholine

CAS Number: 18194-24-6 MDL Number: MFCD00036901 Molecular Formula: C36H72NO8P Molecular Weight: 677.94 Purity/Analysis Method: 97.0% (HPLC,T) Form: Crystal Melting point (°C): 220 Specific rotation [a]20/D: 8.5 deg (C=2, CHCl3) Storage Temperature: 0°C


Product Categories: Bioscience, Lipids, Drug Delivery Systems (DDS) Research Reagents Other Product Names / Synonyms: 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DMPC

Formula: C₃₆H₇₂NO₈P
MW: 677.94 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00036901
CAS Number: 18194-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83f54d2f00a5eef622f9767297776a997c38ce81&vid=83f54d2f00a5eef622f9767297776a997c38ce81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR