Di-tert-butyl pyrocarbonate ∼30% in THF

Supplier: TCI America

Synonyms: Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC

D3879-100ML D3879-500ML
TCD3879-100MLEA 66.9 USD
TCD3879-100ML TCD3879-500ML
Di-tert-butyl pyrocarbonate ∼30% in THF
Di-tert-butyl pyrocarbonate

CAS Number: 24424-99-5
MDL Number: MFCD00008805
Molecular Formula: C10H18O5
Molecular Weight: 218.25
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Di-tert-butyl Pyrocarbonate, Pyrocarbonic Acid Di-tert-butyl Ester, Boc2O

Formula: ((H₃C)₃COC(O))₂O
MDL Number: MFCD00008805
CAS Number: 24424-99-5
UN: 2930
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8191cde168e578e1813a4f16f8b7c5574c52250c&vid=efbadca90798d728c70ca76e3d52abe69ec72da9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR