Dichloromethane-[D₂] (99.9% D)

Supplier: TCI America

Synonyms: Dichloromethane-[²H₂]Dideuteromethylene chlorideMethylene chloride-D2Methylene chloride-d2Dichloromethane-d2

D3529-10ML
TCD3529-10MLEA 186.56 USD
TCD3529-10ML
Dichloromethane-[D₂] (99.9% D)
Dichloromethane-[D2]

CAS Number: 1665-00-5
MDL Number: MFCD00000882
Molecular Formula: CCl2D2
Molecular Weight: 86.94
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 40
Melting point (°C): -97


Other Product Names / Synonyms: Dideuteromethylene Chloride, Methylene Chloride-d2

Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40 °C
Melting Pt: –97 °C
MDL Number: MFCD00000882
CAS Number: 1665-00-5
UN: 1593
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR