3,5-Difluorobenzylamine ≥98.0%

Supplier: TCI America

D3343-1G D3343-5G
TCD3343-1GEA 30 USD
TCD3343-1G TCD3343-5G
3,5-Difluorobenzylamine ≥98.0%
3,5-Difluorobenzylamine

CAS Number: 90390-27-5 MDL Number: MFCD00061244 Molecular Formula: C7H7F2N Molecular Weight: 143.14 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 184 Flash Point (°C): 74 Specific Gravity (20/20): 1.22

Formula: C₇H₇F₂N
Boiling Pt: 184 °C
Density: 1.22 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00061244
CAS Number: 90390-27-5
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eca3a5e9fb5880bcdaadaf17a71cbcfd9f262c4&vid=4eca3a5e9fb5880bcdaadaf17a71cbcfd9f262c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR