N-Ethyldiisopropylamine ∼10% in DMF
Supplier: TCI America
Synonyms:
N,N-Diisopropylethylamine
D2937-10ML
TCD2937-10MLEA
82.22
USD
TCD2937-10ML
N-Ethyldiisopropylamine ∼10% in DMF
N-Ethyldiisopropylamine
[for Detection of Primary and Secondary Amines]
CAS Number: 7087-68-5
MDL Number: MFCD00008868
Molecular Formula: C8H19N
Molecular Weight: 129.25
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: N-Ethyldiisopropylamine, DIPEA, Hunig's Base, DIEA
Formula:
[(CH₃)₂CH]₂NC₂H₅ MW: 129.25 g/mol Density: 0.92 (20 °C) |
MDL Number:
MFCD00008868 CAS Number: 7087-68-5 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...