α,2,6-Trichlorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dichlorobenzyl Chloride1,3-Dichloro-2-(chloromethyl)benzene2,6-Dichlorobenzyl chloride1-(Chloromethyl)-2,6-dichlorobenzene

D1750-25G
TCD1750-25GEA 96.29 USD
TCD1750-25G
α,2,6-Trichlorotoluene ≥98.0% (by GC)
α,2,6-Trichlorotoluene

CAS Number: 2014-83-7
MDL Number: MFCD00000897
Molecular Formula: C7H5Cl3
Molecular Weight: 195.47
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 119
Melting point (°C): 38
Flash Point (°C): 112


Other Product Names / Synonyms: alpha,2,6-Trichlorotoluene

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 119 °C
Melting Pt: 38 °C
Flash Pt: 112 °C
MDL Number: MFCD00000897
CAS Number: 2014-83-7
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/85/31/28978531.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR