N,N'-Dimethylethylendiamine ≥97.0%

Supplier: TCI America

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

D0720-25ML D0720-5ML
TCD0720-25MLEA 159 USD
TCD0720-25ML TCD0720-5ML
N,N'-Dimethylethylendiamine ≥97.0%
N,N'-Dimethylethylendiamine

CAS Number: 110-70-3 MDL Number: MFCD00008290 Molecular Formula: C4H12N2 Molecular Weight: 88.15 Purity/Analysis Method: 97.0% (GC,T) Form: Clear Liquid Boiling point (°C): 119 Flash Point (°C): 26 Specific Gravity (20/20): 0.82


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Ligands Other Product Names / Synonyms: 1,2-Bis(methylamino)ethane, DMEDA

Formula: C₄H₁₂N₂
Boiling Pt: 119 °C
Density: 0.82 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00008290
CAS Number: 110-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&vid=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR