2,3-Dimercapto-1-propanol ≥95.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,3-Dimercaptopropanol
, Dithioglycerine
, Dimercaprol
, Dimercapto-2,3 propanol-1
, 2,3-Dimercaptopropane-1-ol
, 2,3-Dimercaptopropan-1-ol
, BAL
CAS Number: 59-52-9
MDL Number: MFCD00004864
Molecular Formula: C3H8OS2
Molecular Weight: 124.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.24
Other Product Names / Synonyms: Dimercaprol, BAL
Formula:
C₃H₈OS₂ MW: 124.22 g/mol Boiling Pt: 120 °C Density: 1.24 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004864 CAS Number: 59-52-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...