α,α'-Dibromo-m-xylene ≥97.0% (by GC)
Supplier: TCI America
D0215-25G
D0215-5G
TCD0215-25GEA
208.19
USD
TCD0215-25G
TCD0215-5G
α,α'-Dibromo-m-xylene ≥97.0% (by GC)
α,α'-Dibromo-m-xylene
CAS Number: 626-15-3
MDL Number: MFCD00000178
Molecular Formula: C8H8Br2
Molecular Weight: 263.96
Purity/Analysis Method:
Other Product Names / Synonyms: m-Xylylene Bromide, m-Xylylene Dibromide, 1,3-Bis(bromomethyl)benzene
Formula:
C₈H₈Br₂ Boiling Pt: 140 °C Melting Pt: 76 °C |
MDL Number:
MFCD00000178 CAS Number: 626-15-3 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...