ω,4-Dibromoacetophenone ≥98.0% (by GC)

Supplier: TCI America

D0164-25G D0164-100G
TCD0164-25GEA 38.38 USD
TCD0164-25G TCD0164-100G
ω,4-Dibromoacetophenone ≥98.0% (by GC)
ω,4-Dibromoacetophenone

CAS Number: 99-73-0 MDL Number: MFCD00000200 Molecular Formula: C8H6Br2O Molecular Weight: 277.94 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 111 Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,4'-Dibromoacetophenone

Formula: C₈H₆Br₂O
Melting Pt: 111 °C
MDL Number: MFCD00000200
CAS Number: 99-73-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=572ce097b6aa1d8ad12e47dd8f6c3c407e652875&vid=572ce097b6aa1d8ad12e47dd8f6c3c407e652875&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR