3-(Carbamimidoylthio)-1-propanesulfonic acid ≥98.0%
Supplier: TCI America
Synonyms:
3-(Carbamimidoylthio)propane-1-sulphonic acid, 3-(Amidinothio)-1-propanesulfonic acid, 3-(Amidinothio)-1-propanesulphonic acid, 3-(Carbamimidoylthio)propane-1-sulfonic acid
C2982-100G
C2982-25G
TCC2982-100GEA
212.4
USD
TCC2982-100G
TCC2982-25G
3-(Carbamimidoylthio)-1-propanesulfonic acid ≥98.0%
3-(Carbamimidoylthio)-1-propanesulfonic acid
CAS Number: 21668-81-5
MDL Number: MFCD00085942
Molecular Formula: C4H10N2O3S2
Molecular Weight: 198.26
Purity/Analysis Method:
Other Product Names / Synonyms: 3-(Amidinothio)-1-propanesulfonic Acid
Formula:
C₄H₁₀N₂O₃S₂ MW: 198.26 g/mol Melting Pt: 257 °C |
MDL Number:
MFCD00085942 CAS Number: 21668-81-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...