Ciprofloxacin ≥98.0%
Supplier: TCI America
Synonyms:
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
CAS Number: 85721-33-1
MDL Number: MFCD00185755
Molecular Formula: C17H18FN3O3
Molecular Weight: 331.35
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
Formula:
C₁₇H₁₈FN₃O₃ MW: 331.35 g/mol Melting Pt: 265 °C |
MDL Number:
MFCD00185755 CAS Number: 85721-33-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...