Crystal violet for biochemical research
Supplier: TCI America
Synonyms:
4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride
C2460-5G
TCC2460-5GEA
49.77
USD
TCC2460-5G
Crystal violet for biochemical research
Crystal Violet
[for Biochemical Research]
CAS Number: 548-62-9
MDL Number: MFCD00011750
Molecular Formula: C25H30ClN3
Molecular Weight: 407.99
Form: Crystal
Color: Yellow Green
Melting point (°C): 211
Other Product Names / Synonyms: Basic Violet 3, Hexamethylpararosaniline Chloride
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol Melting Pt: 211 °C |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...