Chlorobenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Benzene Chloride
, Phenylchloride
, Monochlorobenzene
C1948-500G
TCC1948-500GEA
23
USD
TCC1948-500G
Chlorobenzene ≥98.0% (by GC)
Chlorobenzene
CAS Number: 108-90-7
MDL Number: MFCD00000530
Molecular Formula: C6H5Cl
Molecular Weight: 112.56
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 132
Melting point (°C): -45
Flash Point (°C): 28
Specific Gravity (20/20): 1.11
Formula:
C₆H₅Cl MW: 112.56 g/mol Boiling Pt: 132 °C Melting Pt: –45 °C Density: 1.11 (20 °C) Flash Pt: 28 °C |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 UN: 1134 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...