Cyclopropylamine ≥95.0% (by GC, titration analysis)
Supplier: TCI America
C0981-25ML
C0981-5ML
TCC0981-5MLEA
20.29
USD
TCC0981-5ML
TCC0981-25ML
Cyclopropylamine ≥95.0% (by GC, titration analysis)
Cyclopropylamine
CAS Number: 765-30-0
MDL Number: MFCD00001301
Molecular Formula: C3H7N
Molecular Weight: 57.10
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 50
Melting point (°C): -10
Flash Point (°C): 1
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: Aminocyclopropane
Formula:
C₃H₇N MW: 57.1 g/mol Boiling Pt: 50 °C Melting Pt: –10 °C Density: 0.82 (20 °C) Flash Pt: 1 °C |
MDL Number:
MFCD00001301 CAS Number: 765-30-0 UN: 2733 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...