Bromocyclobutane ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Cyclobutyl bromide
C0771-1G
C0771-5G
TCC0771-1GEA
34
USD
TCC0771-1G
TCC0771-5G
Bromocyclobutane ≥96.0% (by GC)
Bromocyclobutane
CAS Number: 4399-47-7
MDL Number: MFCD00001317
Molecular Formula: C4H7Br
Molecular Weight: 135.00
Purity/Analysis Method:
Other Product Names / Synonyms: Cyclobutyl Bromide
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 108 °C Density: 1.43 (20 °C) Flash Pt: 22 °C |
MDL Number:
MFCD00001317 CAS Number: 4399-47-7 UN: 1993 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...