Carbachol ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Carbamylcholine chloride

C0596-25G C0596-10G
TCC0596-10GEA 141.92 USD
TCC0596-10G TCC0596-25G
Carbachol ≥99.0% (by titrimetric analysis)
Carbachol

CAS Number: 51-83-2
MDL Number: MFCD00012011
Molecular Formula: C6H15ClN2O2
Molecular Weight: 182.65
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 189


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Reagents for Other Disease Models, Acetylcholine Receptor (AChR) Agent Ingredients
Other Product Names / Synonyms: Carbachol, (2-Carbamoyloxyethyl)trimethylammonium Chloride, Choline Chloride Carbamate

Formula: H₂NC(O)O(CH₂)₂N(CH₃)₃Cl
MW: 182.65 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00012011
CAS Number: 51-83-2
UN: 2811
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/76/33/28937633.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR