2-Bromo-5-methoxybenzyl bromide ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene

B4423-25G B4423-5G
TCB4423-25GEA 222 USD
TCB4423-25G TCB4423-5G
2-Bromo-5-methoxybenzyl bromide ≥98.0%
2-Bromo-5-methoxybenzyl bromide

CAS Number: 19614-12-1 MDL Number: MFCD00078664 Molecular Formula: C8H8Br2O Molecular Weight: 279.96 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 93


Other Product Names / Synonyms: 4-Bromo-3-(bromomethyl)anisole

Formula: C₈H₈Br₂O
MW: 279.96 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00078664
CAS Number: 19614-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de9a71f785d20b0b6a1b772b07a497493348400a&vid=de9a71f785d20b0b6a1b772b07a497493348400a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR