1-Benzyl-N-carbobenzoxy-L-glutamate ≥98.0%
Supplier: TCI America
Synonyms:
N-Cbz-L-glutamic Acid 1-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, Z-Glu-OBzl, 1-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester
CAS Number: 3705-42-8
MDL Number: MFCD00077013
Molecular Formula: C20H21NO6
Molecular Weight: 371.39
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester, 1-Benzyl N-Cbz-L-glutamate, N-Cbz-L-glutamic Acid 1-Benzyl Ester, Z-Glu-OBzl
Formula:
C₂₀H₂₁NO₆ Melting Pt: 100 °C |
MDL Number:
MFCD00077013 CAS Number: 3705-42-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...