4-Bromo-3-fluorobenzenesulfonyl chloride ≥98.0%

Supplier: TCI America

B3835-1G B3835-5G
TCB3835-1GEA 107 USD
TCB3835-1G TCB3835-5G
4-Bromo-3-fluorobenzenesulfonyl chloride ≥98.0%
4-Bromo-3-fluorobenzenesulphonyl chloride

CAS Number: 351003-51-5 MDL Number: MFCD03094150 Molecular Formula: C6H3BrClFO2S Molecular Weight: 273.50 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 218 Flash Point (°C): 110 Specific Gravity (20/20): 1.85

Formula: C₆H₃BrClFO₂S
Boiling Pt: 218 °C
Density: 1.85 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD03094150
CAS Number: 351003-51-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=279c9a0a005d36d6da46fd3076e9f8c9998c4418&vid=279c9a0a005d36d6da46fd3076e9f8c9998c4418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR