1-Bromo-4-ethynylbenzene 98,0
Supplier: TCI America
Synonyms:
4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne
B3701-1G
B3701-5G
TCB3701-1GEA
49
USD
TCB3701-1G
TCB3701-5G
1-Bromo-4-ethynylbenzene 98,0
1-Bromo-4-ethynylbenzene
CAS Number: 766-96-1
MDL Number: MFCD00168822
Molecular Formula: C8H5Br
Molecular Weight: 181.03
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 4-Bromophenylacetylene
Formula:
C₈H₅Br MW: 181.03 g/mol Boiling Pt: 92 °C Melting Pt: 66 °C |
MDL Number:
MFCD00168822 CAS Number: 766-96-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...