Benzyl-4-oxo-1-piperidinecarboxylate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Benzyloxycarbonyl-4-piperidine
, 1-Carbobenzoxy-4-piperidone
, 1-Cbz-4-piperidone
, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester
, 1-Cbz-Piperidin-4-one
CAS Number: 19099-93-5
MDL Number: MFCD00673144
Molecular Formula: C13H15NO3
Molecular Weight: 233.27
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 45
Flash Point (°C): 113
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine
Formula:
C₁₃H₁₅NO₃ MW: 233.27 g/mol Boiling Pt: 170 °C Melting Pt: 45 °C Flash Pt: 113 °C |
MDL Number:
MFCD00673144 CAS Number: 19099-93-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...