X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: X-Gluc

B3620-100MG B3620-10MG
TCB3620-10MGEA 34.17 USD
TCB3620-10MG TCB3620-100MG
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) ≥98.0% (by HPLC) for biochemical research
X-GlcA ((5-Bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)

[for Biochemical Research]
CAS Number: 114162-64-0
MDL Number: MFCD00058543
Molecular Formula: C14H13BrClNO7
Molecular Weight: 521.79
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Cyclohexylammonium Salt, X-Gluc CHA Salt, X-GlcA CHA Salt

Formula: C₂₀H₂₆BrClN₂O₇
MW: 521.79 g/mol
MDL Number: MFCD00058543
CAS Number: 114162-64-0
https://us.vwr.com/stibo/web/std.lang.all/49/47/28934947.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR