1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥97.0% (by HPLC)

Supplier: TCI America

B3428-1G
TCB3428-1GEA 142.81 USD
TCB3428-1G
1,2-Bis(di(pentafluorophenyl)phosphino)ethane ≥97.0% (by HPLC)
1,2-Bis(di(pentafluorophenyl)phosphino)ethane

CAS Number: 76858-94-1
MDL Number: MFCD00058841
Molecular Formula: C26H4F20P2
Molecular Weight: 758.23
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Melting point (°C): 195

Formula: C₂₆H₄F₂₀P₂
Melting Pt: 195 °C
MDL Number: MFCD00058841
CAS Number: 76858-94-1
UN: 3464
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/47/99/28934799.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR