(+)-Bicuculline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 485-49-4
MDL Number: MFCD00005006
Molecular Formula: C20H17NO6
Molecular Weight: 367.36
Purity/Analysis Method:
Formula:
C₂₀H₁₇NO₆ Melting Pt: 197 °C |
MDL Number:
MFCD00005006 CAS Number: 485-49-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...