Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0%

Supplier: TCI America

Synonyms: BOP-Cl

B1213-25G B1213-5G
TCB1213-25GEA 134.84 USD
TCB1213-25G TCB1213-5G
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97.0%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride

CAS Number: 68641-49-6 MDL Number: MFCD00010077 Molecular Formula: C6H8ClN2O5P Molecular Weight: 254.56 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 191


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis Other Product Names / Synonyms: Bis(2-oxo-3-oxazolidinyl)phosphorodiamidic Chloride, BOP-Cl

Formula: C₆H₈ClN₂O₅P
MW: 254.6 g/mol
Melting Pt: 191 °C
MDL Number: MFCD00010077
CAS Number: 68641-49-6
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48887a7481caf4fec003fc0e9f33fdd890f0bb8f&vid=48887a7481caf4fec003fc0e9f33fdd890f0bb8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR