Bis(trimethylsilyl)acetylene ≥97.0%
Supplier: TCI America
B1090-25G
B1090-5G
TCB1090-25GEA
273
USD
TCB1090-25G
TCB1090-5G
Bis(trimethylsilyl)acetylene ≥97.0%
Bis(trimethylsilyl)acetylene
CAS Number: 14630-40-1
MDL Number: MFCD00008276
Molecular Formula: C8H18Si2
Molecular Weight: 170.40
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Click Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: BTMSA
Formula:
C₈H₁₈Si₂ Boiling Pt: 134 °C Flash Pt: 2 °C |
MDL Number:
MFCD00008276 CAS Number: 14630-40-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...