Bromotrimethylsilane ≥95.0% (by GC)
Supplier: TCI America
CAS Number: 2857-97-8
MDL Number: MFCD00000048
Molecular Formula: C3H9BrSi
Molecular Weight: 153.09
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Trimethylbromosilane, Trimethylsilyl Bromide
Formula:
C₃H₉BrSi Boiling Pt: 79 °C Melting Pt: –43 °C Density: 1.18 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 UN: 2924 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...