1-Bromo-3-methyl-2-butene ≥90.0% (by GC) stabilized

Supplier: TCI America

B0844-25G B0844-5G
TCB0844-25GEA 91 USD
TCB0844-25G TCB0844-5G
1-Bromo-3-methyl-2-butene ≥90.0% (by GC) stabilized
1-Bromo-3-methyl-2-butene

(stabilized with Silver chip) CAS Number: 870-63-3 MDL Number: MFCD00000242 Molecular Formula: C5H9Br Molecular Weight: 149.03 Purity/Analysis Method: 90.0% (GC) Form: Clear Liquid Boiling point (°C): 135 Flash Point (°C): 40 Specific Gravity (20/20): 1.29 Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Methyl-2-butenyl Bromide, gamma-Methylcrotyl Bromide

Formula: C₅H₉Br
Boiling Pt: 135 °C
Density: 1.29 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00000242
CAS Number: 870-63-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=924d08ba71465fc1a1529cf62f37fc0a70371706&vid=c478454e3ce1e9818ec7ed54fefd271ee6f65508&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR