Benzene-[D₆] (99.6% D)

Supplier: TCI America

Synonyms: Benzene-[²H₆]

B0840-25ML
TCB0840-25MLEA 258.21 USD
TCB0840-25ML
Benzene-[D₆] (99.6% D)
Benzene-[D6]

CAS Number: 1076-43-3
MDL Number: MFCD00003010
Molecular Formula: C6D6
Molecular Weight: 84.15
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Melting point (°C): 7
Flash Point (°C): -11
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Hexadeuterobenzene

Formula: C₆D₆
MW: 84.07 g/mol
Boiling Pt: 80 °C
Melting Pt: 7 °C
Density: 0.95 (20 °C)
Flash Pt: –11 °C
MDL Number: MFCD00003010
CAS Number: 1076-43-3
UN: 1114
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR