Bromocresol green-methyl red in ethanol mixed indicator
Supplier: TCI America
B0120-100ML
B0120-500ML
TCB0120-100MLEA
58.72
USD
TCB0120-100ML
TCB0120-500ML
Bromocresol green-methyl red in ethanol mixed indicator
Bromocresol green-methyl red
[mixed Indicator]
MDL Number: MFCD00134126
Form: Clear Liquid
Color: Red
Flash Point (°C): 13
Form: Clear Liquid
Color: Red
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...