9-(Chloromethyl)anthracene ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A5502-1G A5502-5G
TCA5502-1GEA 30 USD
TCA5502-1G TCA5502-5G
9-(Chloromethyl)anthracene ≥97.0% (by titrimetric analysis)
9-(Chloromethyl)anthracene

[for HPLC Labeling] CAS Number: 24463-19-2 MDL Number: MFCD00001263 Molecular Formula: C15H11Cl Molecular Weight: 226.70 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 140

Formula: C₁₅H₁₁Cl
Melting Pt: 140 °C
MDL Number: MFCD00001263
CAS Number: 24463-19-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f7da07c43220ec5e6b5bea4ad7aa8805459760a&vid=56bccbca3475afcdd36a88ea1252d6749f076c77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR