Acetazolamide ≥98.0%
Supplier: TCI America
Synonyms:
2-Acetamido-5-sulphonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulphonamide, 2-Acetamido-5-sulfonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide
CAS Number: 59-66-5
MDL Number: MFCD00003105
Molecular Formula: C4H6N4O3S2
Molecular Weight: 222.24
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, 2-Acetamido-5-sulfonamido-1,3,4-thiadiazole
Formula:
C₄H₆N₄O₃S₂ MW: 222.24 g/mol Melting Pt: 259 °C |
MDL Number:
MFCD00003105 CAS Number: 59-66-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...