Atorvastatin calcium salt trihydrate ≥98.0%
Supplier: TCI America
Synonyms:
Calcium (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate trihydrate, Atorvastatin Calcium Salt Trihydrate, Atorvastatin hemicalcium salt sesquihydrate, Atorvastatin calcium
A2476-1G
A2476-5G
TCA2476-1GEA
91.65
USD
TCA2476-1G
TCA2476-5G
Atorvastatin calcium salt trihydrate ≥98.0%
Atorvastatin calcium salt trihydrate
CAS Number: 344423-98-9
MDL Number: MFCD09752074
Molecular Formula: C33H35FN2O5
Molecular Weight: 1155.36
Purity/Analysis Method:
Formula:
C₃₃H₃₅FN₂O₅ |
MDL Number:
MFCD09752074 CAS Number: 344423-98-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...