Acemetacin ≥98.0%

Supplier: TCI America

Synonyms: [1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 2-(2-(1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester

A2452-25G A2452-5G A2452-1G
TCA2452-25GEA 185.2 USD
TCA2452-25G TCA2452-5G TCA2452-1G
Acemetacin ≥98.0%
Acemetacin

CAS Number: 53164-05-9 MDL Number: MFCD00151473 Molecular Formula: C21H18ClNO6 Molecular Weight: 415.83 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 152


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Prodrug Ingredients Other Product Names / Synonyms: [1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester

Formula: C₂₁H₁₈ClNO₆
Melting Pt: 152 °C
MDL Number: MFCD00151473
CAS Number: 53164-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a0d10730f5253e3eb3ffb55ddca3c2e03b4ba86&vid=4a0d10730f5253e3eb3ffb55ddca3c2e03b4ba86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR