2-Aminonicotinonitrile ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 24517-64-4
MDL Number: MFCD00128867
Molecular Formula: C6H5N3
Molecular Weight: 119.13
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Aminonicotinonitrile, 2-Amino-3-pyridinecarbonitrile
Formula:
C₆H₅N₃ Melting Pt: 135 °C |
MDL Number:
MFCD00128867 CAS Number: 24517-64-4 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...