Acetonitrile-[D₃] (99.8% D)

Supplier: TCI America

Synonyms: Acetonitrile-d3Cyanomethane-D3Ethannitrile-D3Methyl-d3 cyanideAcetonitrile-[²H₃]Methyl cyanide-D3Trideuteroacetonitrile

A2018-10ML
TCA2018-10MLEA 97.86 USD
TCA2018-10ML
Acetonitrile-[D₃] (99.8% D)
Acetonitrile-[D3]

CAS Number: 2206-26-0
MDL Number: MFCD00001881
Molecular Formula: C2D3N
Molecular Weight: 44.07
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 6
Specific Gravity (20/20): 0.84


Product Categories: Analytical Science, Deuterated Compounds for NMR
Other Product Names / Synonyms: Trideuteroacetonitrile, Methyl-d3 Cyanide

Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 81 °C
Density: 0.84 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR