2-Amino-1,3,4-thiadiazole ≥98.0%
Supplier: TCI America
Synonyms:
[1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine
A1060-25G
A1060-5G
TCA1060-25GEA
45
USD
TCA1060-25G
TCA1060-5G
2-Amino-1,3,4-thiadiazole ≥98.0%
2-Amino-1,3,4-thiadiazole
CAS Number: 4005-51-0
MDL Number: MFCD00003107
Molecular Formula: C2H3N3S
Molecular Weight: 101.13
Purity/Analysis Method:
Formula:
C₂H₃N₃S MW: 101.13 Melting Pt: 183 °C |
MDL Number:
MFCD00003107 CAS Number: 4005-51-0 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...