Anti-IgG Goat Antibody

Supplier: Advansta
R-05057-250
490016-830EA 191.45 USD
490016-830
Anti-IgG Goat Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR