Anti-FASLG Mouse Antibody (Biotin) [clone: NOK-1]

Supplier: Tonbo Biosciences
30-9919-U025 30-9919-U100
10141-030EA 79.2 USD
10141-030 10141-142
Anti-FASLG Mouse Antibody (Biotin) [clone: NOK-1]
Antibodies
Type:
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR