Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]

Supplier: Tonbo Biosciences
35-5893-U025 35-5893-U100 35-5893-U500
10051-010EA 61.6 USD
10051-010 10051-012 10141-160
Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]
Antibodies
Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR