Anti-IgG Polyclonal Antibody (Europium 1024)

Supplier: Columbia Biosciences
D17-110-20 D17-110-50 D17-110-1MG
10159-206EA 599.15 USD
10159-206 10159-208 10159-236
Anti-IgG Polyclonal Antibody (Europium 1024)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host:
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR