Anti-CD169 Rat Monoclonal Antibody [clone: 3D6.112]

Supplier: Biolegend
142401 142402
142401-BLEA 131.86 USD
142401-BL 142402-BL
Anti-CD169 Rat Monoclonal Antibody [clone: 3D6.112]
Antibodies
Purified anti-mouse CD169 (Siglec-1) [3D6.112]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD169
Clonality: Monoclonal
Clone: 3D6.112
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR