Anti-FOXP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 150D]

Supplier: Biolegend
320007 320008
320007-BLEA 272.88 USD
320007-BL 320008-BL
Anti-FOXP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 150D]
Antibodies
PE anti-mouse/rat/human FOXP3 [150D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat, Cross-Reactivity: Cynomolgus, Rhesus, Baboon; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 150D
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse;Rat;Cross-Reactivity: Cynomolgus;Rhesus;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR