Anti-IgG2a Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMG2a-62]

Supplier: Biolegend
407107 407108
407107-BLEA 94.14 USD
407107-BL 407108-BL
Anti-IgG2a Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMG2a-62]
Antibodies
PE anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG2a
Clonality: Monoclonal
Clone: RMG2a-62
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR