Anti-ZAP-70 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1E7.2]

Supplier: Biolegend
313403 313404
313403-BLEA 131.7 USD
313403-BL 313404-BL
Anti-ZAP-70 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1E7.2]
Antibodies
PE anti-human/mouse ZAP-70 [1E7.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: ZAP-70
Clonality: Monoclonal
Clone: 1E7.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR