Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TER-119]

Supplier: Biolegend
116231 116232
116231-BLEA 148.7 USD
116231-BL 116232-BL
Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: TER-119]
Antibodies
Pacific Blue™ anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.

Type: Primary
Antigen: TER-119/Erythroid Cells
Clonality: Monoclonal
Clone: TER-119
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR