Anti-Cutaneous Lymphocyte Antigen (CLA) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: HECA-452]

Supplier: Biolegend
321307 321308
321307-BLEA 139.6 USD
321307-BL 321308-BL
Anti-Cutaneous Lymphocyte Antigen (CLA) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: HECA-452]
Antibodies
Pacific Blue™ anti-human/mouse Cutaneous Lymphocyte Antigen (CLA) [HECA-452]; Isotype: Rat IgM, κ; Reactivity: Human, Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.

Type: Primary
Antigen: Cutaneous Lymphocyte Antigen (CLA)
Clonality: Monoclonal
Clone: HECA-452
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR