Anti-CD15 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: W6D3]

Supplier: Biolegend
323021 323022 323024
323021-BLEA 158.8 USD
323021-BL 323022-BL 323024-BL
Anti-CD15 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: W6D3]
Antibodies
Pacific Blue™ anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test sizes, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume.
For μg size, the suggested use of this reagent for immunofluorescent staining is 1.0 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: W6D3
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR